3-Amino-N-cyclopropylbenzenesulphonamide 97%

Leverantör: Apollo Scientific

OR59786-1G OR59786-5G
APOSOR59786-1GEA 1200 SEK
APOSOR59786-1G APOSOR59786-5G
3-Amino-N-cyclopropylbenzenesulphonamide 97%
3-Amino-N-cyclopropylbenzenesulphonamide
Formel: C₉H₁₂N₂O₂S
Storage Temperature: Kylskåp
MDL Number: MFCD07022080
CAS nummer: 459434-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR