2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
Leverantör: Apollo Scientific
Varning
OR59471-1G OR59471-250MG OR59471-500MG
APOSOR59471-1GEA 0 SEK
APOSOR59471-1G APOSOR59471-250MG APOSOR59471-500MG
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
2-Amino-5-bromo-3-(hydroxymethyl)pyridine hydrobromide
Formel: C₆H₈Br₂N₂O
Storage Temperature: Omgivande
CAS nummer: 443956-55-6