N-tert-Butylbenzenesulphonamide

Leverantör: Apollo Scientific
Varning

OR59352-1G OR59352-25G OR59352-5G
APOSOR59352-1GEA 790 SEK
APOSOR59352-1G APOSOR59352-25G APOSOR59352-5G
N-tert-Butylbenzenesulphonamide
N-tert-Butylbenzenesulphonamide
Formel: C₁₀H₁₅NO₂S
MW: 213,2966 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00462465
CAS nummer: 2512-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR