1-Benzyl-4-bromo-1H-pyrazole

Leverantör: Apollo Scientific
Varning

OR59333-1G OR59333-5G
APOSOR59333-1GEA 650 SEK
APOSOR59333-1G APOSOR59333-5G
1-Benzyl-4-bromo-1H-pyrazole
1-Benzyl-4-bromo-1H-pyrazole
Formel: C₁₀H₉BrN₂
MW: 237,1000 g/mol
Smältpunkt: 50…54 °C
Storage Temperature: Omgivande
MDL Number: MFCD02179566
CAS nummer: 50877-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR