D-(-)-N-Methylglucamine
Leverantör: Apollo Scientific
Varning
Synonyms: Meglumine, (-)-1-Deoxy-1-(methylamino)-D-glucitol, N-Methyl-D-glucamine, (-)-N-Methyl-D-glucamine, (2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol, (-)-1-Methylamino-1-deoxy-D-glucitol
OR5855-250G OR5855-50G
APOSOR5855-250GEA 685 SEK
APOSOR5855-250G APOSOR5855-50G
D-(-)-N-Methylglucamine
D-(-)-N-Methylglucamine
Formel: H₃CNHCH₂(CH(OH))₄CH₂OH
MW: 195,22 g/mol
Smältpunkt: 129…131,5 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004707
CAS nummer: 6284-40-8
EINECS: 228-506-9