4-Phenylbenzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
OR5838-100G OR5838-500G
APOSOR5838-100GEA 380 SEK
APOSOR5838-100G APOSOR5838-500G
4-Phenylbenzoic acid 98%
4-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Smältpunkt: 224…226 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002553
CAS nummer: 92-92-2
EINECS: 202-203-1