3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid 95%

Leverantör: Apollo Scientific

Synonyms: 3-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(pyrrolidinyl-1-carbonyl)phenylboronic acid

OR5813-1G
APOSOR5813-1GEA 3550 SEK
APOSOR5813-1G
3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid 95%
3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid
Formel: C₁₁H₁₃BClNO₃
MW: 253.49 g/mol
Smältpunkt: 152…155 °C
Storage Temperature: Omgivande
MDL Number: MFCD07363776
CAS nummer: 850589-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR