3-Acetylthiophene 98%

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 3-thienyl ketone, 1-(3-Thienyl)-1-ethanone

OR5739-1G OR5739-5G
APOSOR5739-1GEA 245 SEK
APOSOR5739-1G APOSOR5739-5G
3-Acetylthiophene 98%
3-Acetylthiophene
Formel: C₆H₆OS
MW: 126,18 g/mol
Kokpunkt: 208…210 °C/748mmHg
Smältpunkt: 57…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005468
CAS nummer: 1468-83-3
EINECS: 215-996-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR