3-Bromo-N-butylbenzenesulphonamide 98%

Leverantör: Apollo Scientific

OR5728-1G OR5728-5G
APOSOR5728-1GEA 1475 SEK
APOSOR5728-1G APOSOR5728-5G
3-Bromo-N-butylbenzenesulphonamide 98%
3-Bromo-N-butylbenzenesulphonamide
Formel: C₁₀H₁₄BrNO₂S
Smältpunkt: 46…50 °C
Storage Temperature: Kylskåp
MDL Number: MFCD07363822
CAS nummer: 871269-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR