3-Bromo-N,N-dimethylbenzenesulphonamide 98%

Leverantör: Apollo Scientific
Varning

OR5726-1G OR5726-25G OR5726-5G
APOSOR5726-1GEA 795 SEK
APOSOR5726-1G APOSOR5726-25G APOSOR5726-5G
3-Bromo-N,N-dimethylbenzenesulphonamide 98%
3-Bromo-N,N-dimethylbenzenesulphonamide
Formel: C₈H₁₀BrNO₂S
Smältpunkt: 74…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD07363826
CAS nummer: 153435-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR