4-Bromo-N-cyclohexylbenzenesulphonamide 98%

Leverantör: Apollo Scientific

OR5706-10G OR5706-1G OR5706-5G
APOSOR5706-10GEA 6450 SEK
APOSOR5706-10G APOSOR5706-1G APOSOR5706-5G
4-Bromo-N-cyclohexylbenzenesulphonamide 98%
4-Bromo-N-cyclohexylbenzenesulphonamide
Formel: C₁₂H₁₆BrNO₂S
Smältpunkt: 102…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00450584
CAS nummer: 7454-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR