3-Acetylphenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3'-Boronoacetophenone, 3-Acetylphenylboronic Acid, 3-Acetylbenzeneboronic acid, Dihydroxy(3-acetylphenyl)borane, (3-Acetylphenyl)metaboric acid, 3-Acetylphenylboronic acid, 3-Acetylbenzeneboronic acid
OR5678-100G OR5678-25G OR5678-5G
APOSOR5678-100GEA 2750 SEK
APOSOR5678-100G APOSOR5678-25G APOSOR5678-5G
3-Acetylphenylboronic acid
3-Acetylphenylboronic acid
Formel: C₈H₉BO₃
MW: 163.97
Smältpunkt: 204…208 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01074678
CAS nummer: 204841-19-0