5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 97%
Leverantör: Apollo Scientific
Varning
OR55705-1G OR55705-250MG
APOSOR55705-1GEA 2425 SEK
APOSOR55705-1G APOSOR55705-250MG
5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine 97%
5,6,7,8-Tetrahydro[1,2,4]triazolo[1,5-a]pyrazine
Formel: C₅H₈N₄
MW: 124,1400 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09835083
CAS nummer: 233278-56-3