Methyl-3-iodo-2-methylbenzoate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 3-iodo-2-methylbenzoate
OR55477-1G OR55477-5G OR55477-10G
APOSOR55477-1GEA 600 SEK
APOSOR55477-1G APOSOR55477-5G APOSOR55477-10G
Methyl-3-iodo-2-methylbenzoate 98%
Methyl-3-iodo-2-methylbenzoate
Formel: C₉H₉IO₂
MW: 276,0710 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD12923250
CAS nummer: 52570-33-9