(S)-Methyl-2,2-dimethyl-1,3-dioxolane-4-carboxylate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-Methyl 2,2-dimethyl-1,3-dioxolane-4-carboxylate
OR55409-1G OR55409-25G OR55409-5G
APOSOR55409-1GEA 945 SEK
APOSOR55409-1G APOSOR55409-25G APOSOR55409-5G
(S)-Methyl-2,2-dimethyl-1,3-dioxolane-4-carboxylate 97%
(S)-Methyl-2,2-dimethyl-1,3-dioxolane-4-carboxylate
Formel: C₇H₁₂O₄
MW: 160,1700 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00066526
CAS nummer: 60456-21-5