Diglycolic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2,2'-Oxydiacetic acid
OR55403-100G
OR55403-500G
APOSOR55403-100GEA
1625
SEK
APOSOR55403-100G
APOSOR55403-500G
Diglycolic acid 98%
Diglycolic acid
Formel:
C₄H₆O₅ MW: 134,09 g/mol Smältpunkt: 140…145 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00004309 CAS nummer: 110-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...