[4-(Chlorosulphonyl)phenyl]acetic acid, Technical Grade
Leverantör: Apollo Scientific
Fara
OR5536-1G OR5536-5G
APOSOR5536-1GEA 3500 SEK
APOSOR5536-1G APOSOR5536-5G
[4-(Chlorosulphonyl)phenyl]acetic acid, Technical Grade
[4-(Chlorosulphonyl)phenyl]acetic acid
Formel: C₈H₇ClO₄S
MW: 234,66 g/mol
Smältpunkt: 125…136 °C
Storage Temperature: Omgivande
MDL Number: MFCD03094677
CAS nummer: 22958-99-2
UN: 3261
ADR: 8,III