4-Methoxydiphenylamine 97%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
4-Methoxy-N-phenylaniline
OR55248-10G
OR55248-25G
OR55248-5G
APOSOR55248-10GEA
1025
SEK
APOSOR55248-10G
APOSOR55248-25G
APOSOR55248-5G
4-Methoxydiphenylamine 97%
4-Methoxydiphenylamine
Formel:
C₁₃H₁₃NO MW: 199,2500 g/mol Kokpunkt: 195 °C/12mmHg Smältpunkt: 106 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00228649 CAS nummer: 1208-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...