1-(3-sulphopropyl)pyridinium hydroxide inner salt

Leverantör: Apollo Scientific
Varning

Synonyms: NDSB 201, 3-(1-Pyridinio)propanesulphonate, 3-(Pyridin-1-ium-1-yl)propane-1-sulphonate

OR55211-25G OR55211-100G
APOSOR55211-25GEA 895 SEK
APOSOR55211-25G APOSOR55211-100G
1-(3-sulphopropyl)pyridinium hydroxide inner salt
1-(3-sulphopropyl)pyridinium hydroxide inner salt
Formel: C₈H₁₁NO₃S
MW: 201,24 g/mol
Smältpunkt: 279 °C
Storage Temperature: Omgivande
MDL Number: MFCD00064468
CAS nummer: 15471-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR