trans-4-Hydroxycinnamic acid
Leverantör: Apollo Scientific
Varning
Synonyms: p-Coumaric acid, trans-p-Cumaric Acid, trans-p-Coumaric Acid
OR55198-100G OR55198-25G OR55198-5G
APOSOR55198-100GEA 2500 SEK
APOSOR55198-100G APOSOR55198-25G APOSOR55198-5G
trans-4-Hydroxycinnamic acid
trans-4-Hydroxycinnamic acid
Formel: C₉H₈O₃
MW: 164.16 g/mol
Smältpunkt: 214 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004399
CAS nummer: 501-98-4