Diethylene glycol monobenzyl ether 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-[2-(Benzyloxy)ethoxy]ethanol, 2-(2-(Benzyloxy)ethoxy)ethanol
OR55194-100G OR55194-1KG OR55194-500G
APOSOR55194-100GEA 1900 SEK
APOSOR55194-100G APOSOR55194-1KG APOSOR55194-500G
Diethylene glycol monobenzyl ether 98%
Diethylene glycol monobenzyl ether
Formel: C₁₁H₁₆O₃
MW: 196,25 g/mol
Densitet: 1,08 g/cm³ (25 °C)
Flampunkt: 158 °C
Storage Temperature: Omgivande
MDL Number: MFCD00029649
CAS nummer: 2050-25-1