(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
(E)-2-(2-Ethoxyvinyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
OR55131-1G
OR55131-5G
OR55131-25G
OR55131-10G
APOSOR55131-1GEA
0
SEK
APOSOR55131-1G
APOSOR55131-5G
APOSOR55131-25G
APOSOR55131-10G
(E)-1-Ethoxyethene-2-boronic acid pinacolester 98%
(E)-1-Ethoxyethene-2-boronic acid pinacolester
Formel:
C₁₀H₁₉BO₃ Kokpunkt: 50 °C/0,3mmHg Densitet: 0,935 g/cm³ (25 °C) Storage Temperature: Frys |
MDL Number:
MFCD09998813 CAS nummer: 1201905-61-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...