Ethyl-2-chloroacetoacetate

Leverantör: Apollo Scientific
Fara

Synonyms: Ethyl 2-chloroacetoacetate

OR5481-100G OR5481-500G
APOSOR5481-100GEA 1075 SEK
APOSOR5481-100G APOSOR5481-500G
Ethyl-2-chloroacetoacetate
Ethyl-2-chloroacetoacetate
Formel: C₆H₉ClO₃
MW: 164,59 g/mol
Kokpunkt: 107 °C/14mmHg
Smältpunkt: < –80 °C
Densitet: 1,19 g/cm³ (25 °C)
Flampunkt: 54 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009141
CAS nummer: 609-15-4
EINECS: 210-180-4
UN: 2920
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR