N-(2-Oxoethyl)phthalimide 97%

Leverantör: Apollo Scientific
Varning

Synonyms: N-(Formylmethyl)phthalimide, Phthalimidoacetaldehyde

OR54675-25G OR54675-5G
APOSOR54675-25GEA 1150 SEK
APOSOR54675-25G APOSOR54675-5G
N-(2-Oxoethyl)phthalimide 97%
N-(2-Oxoethyl)phthalimide
Formel: C₁₀H₇NO₃
MW: 189,17 g/mol
Smältpunkt: 114 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00023080
CAS nummer: 2913-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR