3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid
OR54619-500G OR54619-100G OR54619-25G
APOSOR54619-500GEA 2450 SEK
APOSOR54619-500G APOSOR54619-100G APOSOR54619-25G
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid 97%
3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid
Formel: C₁₇H₂₆O₃
MW: 278,40 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00017519
CAS nummer: 20170-32-5