2-Cyano-2-propyl Dodecyl trithiocarbonate 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Cyanopropan-2-yl Dodecyl Carbonotrithioate, 2-Cyano-2-propanyl Dodecyl Carbonotrithioate
OR54521-1G OR54521-5G
APOSOR54521-1GEA 3025 SEK
APOSOR54521-1G APOSOR54521-5G
2-Cyano-2-propyl Dodecyl trithiocarbonate 97%
2-Cyano-2-propyl Dodecyl trithiocarbonate
Formel: C₁₇H₃₁NS₃
MW: 345.62 g/mol
Densitet: 0.991 g/cm³ (25 °C)
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD11973800
CAS nummer: 870196-83-1