N-Carbobenzoxy-1,3-diaminopropane hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: N-Benzyloxycarbonyl-1,3-propanediamine hydrochloride, N-Z-1,3-Propanediamine hydrochloride, N-Benzyloxycarbonyl-1,3-diaminopropane hydrochloride, N-Cbz-1,3-diaminopropane Hydrochloride, Benzyl N-(3-Aminopropyl)carbamate Hydrochloride, N-(3-Aminopropyl)carbamic Acid Benzyl Ester Hydrochloride, Benzyl (3-aminopropyl)carbamate hydrochloride
OR54485-1G OR54485-5G
APOSOR54485-1GEA 1125 SEK
APOSOR54485-1G APOSOR54485-5G
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
N-Carbobenzoxy-1,3-diaminopropane hydrochloride
Formel: C₁₁H₁₆N₂O₂
MW: 244,72 g/mol
Smältpunkt: 185…189 °C
Storage Temperature: Omgivande
MDL Number: MFCD00270153
CAS nummer: 17400-34-9