3-(Hydroxymethyl)benzyl alcohol
Leverantör: Apollo Scientific
Varning
Synonyms: 1,3-Benzenedimethanol, m-Xylene-α,α'-diol, m-Phenylenedimethanol
OR54383-25G OR54383-5G OR54383-100G
APOSOR54383-25GEA 990 SEK
APOSOR54383-25G APOSOR54383-5G APOSOR54383-100G
3-(Hydroxymethyl)benzyl alcohol
3-(Hydroxymethyl)benzyl alcohol
Formel: C₈H₁₀O₂
MW: 138.17 g/mol
Kokpunkt: 154…159 °C/13mmHg
Smältpunkt: 56…60 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004649
CAS nummer: 626-18-6
EINECS: 210-934-2