4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
Leverantör: Apollo Scientific
Fara
OR53047-100G OR53047-25G OR53047-5G
APOSOR53047-100GEA 2850 SEK
APOSOR53047-100G APOSOR53047-25G APOSOR53047-5G
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
Formel: C₁₄H₁₇ClN₂O₄
MW: 312,7500 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09751265
CAS nummer: 183322-18-1