3-Bromo-5-(4-morpholino)pyridine 97%
Leverantör: Apollo Scientific
Synonyms: 4-(5-Bromo-3-pyridyl)morpholine, 3-Bromo-5-morpholinopyridine
OR52763-1G OR52763-250MG OR52763-5G
APOSOR52763-1GEA 1900 SEK
APOSOR52763-1G APOSOR52763-250MG APOSOR52763-5G
3-Bromo-5-(4-morpholino)pyridine 97%
3-Bromo-5-(4-morpholino)pyridine
Formel: C₉H₁₁BrN₂O
MW: 243,1004 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11656285
CAS nummer: 200064-13-7