trans-10-Hydroxy-2-decenoic acid 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
(E)-10-Hydroxydec-2-enoic acid, Royal Jelly acid
OR52746-1G
OR52746-5G
APOSOR52746-1GEA
2600
SEK
APOSOR52746-1G
APOSOR52746-5G
trans-10-Hydroxy-2-decenoic acid 98%
trans-10-Hydroxy-2-decenoic acid
Formel:
C₁₀H₁₈O₃ MW: 186,25 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00204506 CAS nummer: 14113-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...