Fmoc-Sar-OH 98%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methylglycine, N-Fmoc-N-methylglycine, Fmoc-N-Me-Gly-OH
OR52600-100G
OR52600-10G
OR52600-25G
APOSOR52600-100GEA
1700
SEK
APOSOR52600-100G
APOSOR52600-10G
APOSOR52600-25G
Fmoc-Sar-OH 98%
Fmoc-Sar-OH
Formel:
C₁₈H₁₇NO₄ Smältpunkt: 115 °C (dec.) Storage Temperature: Kylskåp |
MDL Number:
MFCD00151926 CAS nummer: 77128-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...