(4-Bromopyridin-2-yl)methanamine 95%
Leverantör: Apollo Scientific
Fara
OR52531-1G OR52531-5G
APOSOR52531-1GEA 2350 SEK
APOSOR52531-1G APOSOR52531-5G
(4-Bromopyridin-2-yl)methanamine 95%
(4-Bromopyridin-2-yl)methanamine
Formel: C₆H₇BrN₂
Storage Temperature: Omgivande
MDL Number: MFCD07374903
CAS nummer: 865156-50-9