6-Methyl-3(2H)-pyridazinone
Leverantör: Apollo Scientific
Varning
Synonyms: 6-Methylpyridazin-3(2H)-one
OR52459-25G OR52459-5G
APOSOR52459-25GEA 875 SEK
APOSOR52459-25G APOSOR52459-5G
6-Methyl-3(2H)-pyridazinone
6-Methyl-3(2H)-pyridazinone
Formel: C₅H₆N₂O
MW: 110,12 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00039720
CAS nummer: 13327-27-0