2,8-Diazaspiro[4.5]decan-3-one 95+%
Leverantör: Apollo Scientific
Varning
OR52408-250MG OR52408-1G OR52408-5G
APOSOR52408-250MGEA 875 SEK
APOSOR52408-250MG APOSOR52408-1G APOSOR52408-5G
2,8-Diazaspiro[4.5]decan-3-one 95+%
2,8-Diazaspiro[4.5]decan-3-one
Formel: C₈H₁₄N₂O
Storage Temperature: Kylskåp
MDL Number: MFCD11044206
CAS nummer: 561314-57-6