Etyl-4-aminbensoat 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Ethyl 4-aminobenzoate, Aethoform
OR52189-100G OR52189-25G
APOSOR52189-100GEA 550 SEK
APOSOR52189-100G APOSOR52189-25G
Etyl-4-aminbensoat 98%
Etyl-4-aminbensoat
Formel: C₉H₁₁NO₂
MW: 165,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00007892
CAS nummer: 94-09-7