1-Methyl-1H-pyrazole-5-carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Methyl-5-pyrazolecarboxylic acid
OR5218-1G OR5218-10G OR5218-5G
APOSOR5218-1GEA 175 SEK
APOSOR5218-1G APOSOR5218-10G APOSOR5218-5G
1-Methyl-1H-pyrazole-5-carboxylic acid 97%
1-Methyl-1H-pyrazole-5-carboxylic acid
Formel: C₅H₆N₂O₂
MW: 126,11 g/mol
Smältpunkt: 220…225 °C
Storage Temperature: Omgivande
MDL Number: MFCD00464253
CAS nummer: 16034-46-1