2-Chloroquinoline-5-carboxylic acid 98%

Leverantör: Apollo Scientific
Varning

OR52151-1G OR52151-5G
APOSOR52151-1GEA 1100 SEK
APOSOR52151-1G APOSOR52151-5G
2-Chloroquinoline-5-carboxylic acid 98%
2-Chloroquinoline-5-carboxylic acid
Formel: C₁₀H₆ClNO₂
MW: 207,6100 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11226761
CAS nummer: 1092287-30-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR