Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Leverantör: Apollo Scientific
Varning
Synonyms: trans-Dichlorobis(tricyclohexylphosphine)palladium(II), Dichlorobis(tricyclohexylphosphine)palladium(II)
OR52149-1G OR52149-5G OR52149-10G
APOSOR52149-1GEA 650 SEK
APOSOR52149-1G APOSOR52149-5G APOSOR52149-10G
Bis(tricyclohexylphosphine)palladium(II) dichloride 98%
Bis(tricyclohexylphosphine)palladium(II) dichloride
Formel: C₃₆H₆₆Cl₂P₂Pd
MW: 738,19 g/mol
Smältpunkt: 270 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00191830
CAS nummer: 29934-17-6