4,4'-Bis(2-benzoxazolyl)stilbene
Leverantör: Apollo Scientific
Varning
Synonyms: 1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethene
OR52140-100G OR52140-1KG OR52140-500G
APOSOR52140-100GEA 275 SEK
APOSOR52140-100G APOSOR52140-1KG APOSOR52140-500G
4,4'-Bis(2-benzoxazolyl)stilbene
4,4'-Bis(2-benzoxazolyl)stilbene
Optical brightener for plastics (PP, ABS, EVA, PS and PC) and fibre (polyester fibres and PP fibres).
Formel: C₂₈H₁₈N₂O₂
MW: 414,46 g/mol
Smältpunkt: 353…359 °C
Storage Temperature: Omgivande
MDL Number: MFCD00022846
CAS nummer: 1533-45-5