Ethyl-2-acetyl-3-ethoxyacrylate

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 2-(ethoxymethylene)acetoacetate, Ethyl 2-(ethoxymethylene)-3-oxobutanoate

OR51989-1G OR51989-5G
APOSOR51989-1GEA 635 SEK
APOSOR51989-1G APOSOR51989-5G
Ethyl-2-acetyl-3-ethoxyacrylate
Ethyl-2-acetyl-3-ethoxyacrylate
Formel: C₉H₁₄O₄
MW: 186,21 g/mol
Kokpunkt: 264…266 °C
Densitet: 1,07 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD09959866
CAS nummer: 3788-94-1
EINECS: 223-259-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR