Benzyl nicotinate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Picolinic acid benzyl ester, Pyridine-3-carboxylic acid benzyl ester, Benzyl 3-pyridinecarboxylate, 3-Pyridinecarboxylic acid benzyl ester, Nicotinic acid benzyl ester
OR51903-250G OR51903-50G OR51903-1KG
APOSOR51903-250GEA 1375 SEK
APOSOR51903-250G APOSOR51903-50G APOSOR51903-1KG
Benzyl nicotinate 98%
Benzyl nicotinate
Formel: C₁₃H₁₁NO₂
Kokpunkt: 177 °C/8,25mmHg
Smältpunkt: 24 °C
Densitet: 1,165 g/cm³ (25 °C)
Flampunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00023584
CAS nummer: 94-44-0