EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Leverantör: Apollo Scientific
Fara
Synonyms: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
OR5163-100G OR5163-500G OR5163-1KG
APOSOR5163-100GEA 310 SEK
APOSOR5163-100G APOSOR5163-500G APOSOR5163-1KG
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride)
Formel: C₈H₁₇N₃·HCl
MW: 191.7 g/mol
Smältpunkt: 110…115 °C
Storage Temperature: Frys
MDL Number: MFCD00012503
CAS nummer: 25952-53-8
EINECS: 247-361-2