1-Acetylpiperazine 97%
Leverantör: Apollo Scientific
|
Varning
|
OR5122-100G
OR5122-25G
OR5122-500G
OR5122-5G
APOSOR5122-100GEA
1200
SEK
APOSOR5122-100G
APOSOR5122-25G
APOSOR5122-500G
APOSOR5122-5G
1-Acetylpiperazine 97%
1-Acetylpiperazine
Formel:
C₆H₁₂N₂O MW: 128,17 g/mol Kokpunkt: 127…130 °C/8mmHg Smältpunkt: 31…34 °C Flampunkt: 113 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00058676 CAS nummer: 13889-98-0 EINECS: 237-659-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...