Kloreten 99%

Leverantör: Apollo Scientific

OR51188-25G OR51188-100G
APOSOR51188-25GEA 0 SEK
APOSOR51188-25G APOSOR51188-100G
Kloreten 99%
Kloreten
Formel: C₂H₃Cl
Kokpunkt: –13,4 °C
Densitet: 0,911 g/cm³ (25 °C)
Flampunkt: –61 °C
Storage Temperature: Omgivande
CAS nummer: 75-01-4
UN: 1086
ADR: 2.1,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR