4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 99%
Leverantör: Apollo Scientific
Synonyms:
2-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]triphenylene, 4,4,5,5-Tetramethyl-2-(3-(triphenylen-2-yl)phenyl)-1,3,2-dioxaborolane
OR51013-1G
OR51013-5G
APOSOR51013-1GEA
1400
SEK
APOSOR51013-1G
APOSOR51013-5G
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 99%
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane
Formel:
C₃₀H₂₇BO₂ MW: 430,35 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD28130383 CAS nummer: 1115639-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...