2,1,3-benzoxadiazole-4-sulphonyl chloride, Technical Grade
Leverantör: Apollo Scientific
Fara
Synonyms: Benzofurazan-4-sulphonyl chloride
OR4939-1G OR4939-5G OR4939-25G
APOSOR4939-1GEA 550 SEK
APOSOR4939-1G APOSOR4939-5G APOSOR4939-25G
2,1,3-benzoxadiazole-4-sulphonyl chloride, Technical Grade
2,1,3-benzoxadiazole-4-sulphonyl chloride
Formel: C₆H₃ClN₂O₃S
MW: 218,611 g/mol
Smältpunkt: 77…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00068072
CAS nummer: 114322-14-4
UN: 3261
ADR: 8,II