Di(1-adamantyl)-n-butylphosphine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: n-butyl-di-adamantylphosphine, [cataCXium® A], Bis(1-adamantyl)-butyl-phosphane, Butyldi-1-adamantylphosphine, Di(adamantan-1-yl)(butyl)phosphine
OR49071-1G OR49071-25G OR49071-5G
APOSOR49071-1GEA 585 SEK
APOSOR49071-1G APOSOR49071-25G APOSOR49071-5G
Di(1-adamantyl)-n-butylphosphine 98%
Di(1-adamantyl)-n-butylphosphine
Formel: C₂₄H₃₉P
MW: 358.55 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05861606
CAS nummer: 321921-71-5