2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
Leverantör: Apollo Scientific
OR49043-25G OR49043-50G OR49043-5G
APOSOR49043-25GEA 6425 SEK
APOSOR49043-25G APOSOR49043-50G APOSOR49043-5G
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
2,2-Dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one
Formel: C₉H₁₀N₂O₂
MW: 178,1879 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10000810
CAS nummer: 20348-21-4