2-Hydroxyisobutyric acid
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Hydroxy-2-methylpropionic acid, α-Hydroxyisobutyric acid, 2-Hydroxy-2-methylpropanoic acid
OR4840-100G OR4840-25G OR4840-500G OR4840-1KG
APOSOR4840-100GEA 325 SEK
APOSOR4840-100G APOSOR4840-25G APOSOR4840-500G APOSOR4840-1KG
2-Hydroxyisobutyric acid
2-Hydroxyisobutyric acid
Formel: C₄H₈O₃
MW: 104,11 g/mol
Kokpunkt: 84 °C/1,5mmHg
Smältpunkt: 76…80 °C
Densitet: 1,24 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004459
CAS nummer: 594-61-6